Merck
A porous tantalum trabecular metal: basic science.
American journal of orthopedics (Belle Mead, N.J.) (2002-05-15)
Robert Cohen
ABSTRACT
Conventional materials used for orthopedic reconstruction implants limit the design of new and improved implants. Solid metal is rigid. Porous fixation surfaces have low volumetric porosity available for biologic ingrowth and low frictional characteristics for initial implant stability and have to be bonded to solid-metal substrates. New porous structural biomaterials have been developed to help advance the current generation of prostheses and address unresolved clinical applications. Tantalum trabecular metal offers several advantages over other current conventional materials used for implants by its uniformity and structural continuity, strength, low stiffness, high porosity, and high coefficient of friction.
Sigma-Aldrich
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, 60-100 mesh, 99.9%
Tantalum, foil, 10mm disks, thickness 0.03mm, as rolled, 99.9%
Tantalum, foil, 0.2m coil, thickness 0.1mm, coil width .62mm, as rolled, 99.9%
Tantalum, foil, 50x50mm, thickness 0.5mm, annealed, 99.9%
Tantalum, foil, 10m coil, thickness 0.03mm, coil width 0.75mm, annealed, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.01mm, 99.9%
Tantalum, foil, 10mm disks, thickness 0.0075mm, 99.9%
Tantalum, foil, 10mm disks, thickness 0.0125mm, 99.9%
Tantalum, foil, 10mm disks, thickness 0.015mm, 99.9%
Tantalum, rod, 50mm, diameter 12.7mm, annealed, 99.9%
Tantalum, foil, 4mm disks, thickness 0.75mm, annealed, 99.9%
Tantalum, foil, 6mm disks, thickness 0.009mm, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.001mm, 99.9%
Tantalum, foil, 8mm disks, thickness 0.25mm, annealed, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.0015mm, 99.9%
Tantalum, tube, 100mm, outside diameter 0.31mm, inside diameter 0.19mm, wall thickness 0.06mm, as drawn, 99.9%
Tantalum, foil, 6mm disks, thickness 0.0125mm, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.0015mm, 99.9%
Tantalum, tube, 100mm, outside diameter 3.0mm, inside diameter 2mm, wall thickness 0.5mm, as drawn, 99.9%
Tantalum, foil, not light tested, 82x100mm, thickness 0.006mm, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.006mm, 99.9%
Tantalum, rod, 50mm, diameter 18mm, annealed, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.015mm, 99.9%
Tantalum, foil, 6mm disks, thickness 0.015mm, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.01mm, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.01mm, 99.9%
Tantalum, foil, 8mm disks, thickness 0.025mm, as rolled, 99.9%
Tantalum, foil, light tested, 100x100mm, thickness 0.034mm, as rolled, 99.9%