Merck
  • Dendritic architectures based on bis-MPA: functional polymeric scaffolds for application-driven research.
Dendritic architectures based on bis-MPA: functional polymeric scaffolds for application-driven research.
Chemical Society reviews (2013-05-01)
Anna Carlmark, Eva Malmström, Michael Malkoch
ABSTRACT
Dendritic polymers are highly branched, globular architectures with multiple representations of functional groups. These nanoscale organic frameworks continue to fascinate researchers worldwide and are today under intensive investigation in application-driven research. A large number of potential application areas have been suggested for dendritic polymers, including theranostics, biosensors, optics, adhesives and coatings. The transition from potential to real applications is strongly dictated by their commercial accessibility, scaffolding ability as well as biocompatibility. A dendritic family that fulfills these requirements is based on the 2,2-bismethylolpropionic acid (bis-MPA) monomer. This critical review is the first of its kind to cover most of the research activities generated on aliphatic polyester dendritic architectures based on bis-MPA. It is apparent that these scaffolds will continue to be in the forefront of cutting-edge research as their structural variations are endless including dendrons, dendrimers, hyperbranched polymers, dendritic-linear hybrids and their hybridization with inorganic surfaces.
Sigma-Aldrich
Bis-MPA-NHBoc dendrimer, generation 4
Sigma-Aldrich
Polyester bis-MPA dendron 2 NHBoc, 1 Biotin (core), generation 1
Sigma-Aldrich
Polyester bis-MPA dendron 8 carboxyl, 1 biotin (core), generation 3
Sigma-Aldrich
Polyester bis-MPA dendron 2 NHBoc, 1 azide (core), generation 1
Sigma-Aldrich
Poly(ethylene glycol) linear dendrimer, NHBoc terminated, PEG Mn 6000, generation 4
Sigma-Aldrich
Poly(ethylene glycol) linear dendrimer, carboxyl terminated, PEG Mn 10K, generation 5
Sigma-Aldrich
Polyester bis-MPA dendron, 2 carboxyl, 1 biotin (core), generation 1
Sigma-Aldrich
Poly(ethylene glycol) linear dendron, carboxyl terminated, PEG Mn 10,000, generation 1
Sigma-Aldrich
Poly(ethylene glycol) linear dendrimer, carboxyl terminated, PEG Mn 10k, generation 4
Sigma-Aldrich
Poly(ethylene glycol) linear dendron, carboxyl terminated, PEG Mn 10,000, generation 4
Sigma-Aldrich
Polyester bis-MPA dendron 8 NHBoc, 1 azide (core), generation 3
Sigma-Aldrich
Polyester bis-MPA dendron 4 NHBoc, 1 azide (core), generation 2
Sigma-Aldrich
Polyester bis-MPA dendron, no. Surface Groups 4, NHBoc, 1 Biotin (core), generation 2
Sigma-Aldrich
Polyester bis-MPA dendron, 4 Rhodamine, 1 carboxyl (core), generation 2
Sigma-Aldrich
bis-MPA-OH dendrimer, trimethylol propane core, generation 2
Sigma-Aldrich
bis-MPA-COOH dendrimer, trimethylol propane core, generation 1
Sigma-Aldrich
bis-MPA-Azide dendrimer, trimethylol propane core, generation 5
Sigma-Aldrich
bis-MPA-Azide dendrimer, trimethylol propane core, generation 3
Sigma-Aldrich
Polyester bis-MPA dendron, 4 NHBoc, 1 NHS (core), generation 2
Sigma-Aldrich
Bis-MPA-RAFT dendrimer, trimethylol propane core, generation 1
Sigma-Aldrich
Polyester bis-MPA dendron 8 NHBoc, 1 NHS (core), generation 3
Sigma-Aldrich
bis-MPA-Acetylene dendrimer, trimethylol propane core, generation 3
Sigma-Aldrich
Poly(ethylene glycol) linear dendron, carboxyl terminated, PEG Mn 10,000, generation 2
Sigma-Aldrich
Polyester bis-MPA dendron, no. Surface Groups 8, 8 acetylene, 1 NHBoc (core), generation 3
Sigma-Aldrich
Poly(ethylene glycol) linear dendrimer, carboxyl terminated, PEG Mn 10k, generation 3
Sigma-Aldrich
Polyester bis-MPA dendron 2 NHBoc, 1 carboxyl (core), generation 1
Sigma-Aldrich
Polyester bis-MPA dendron, generation 2, no. Surface Groups 4
Sigma-Aldrich
Polyester bis-MPA dendron, 4 carboxyl, 1 Rhodamine (core), generation 2
Sigma-Aldrich
Polyester bis-MPA dendron, 16 acetylene, 1 NHBoc (core), generation 4
Sigma-Aldrich
bis-MPA-COOH dendrimer, trimethylol propane core, generation 4