Merck
  • The effect of Ti anodized nano-foveolae structure on preosteoblast growth and osteogenic gene expression.
The effect of Ti anodized nano-foveolae structure on preosteoblast growth and osteogenic gene expression.
Journal of nanoscience and nanotechnology (2014-04-18)
Wei-Qiang Yu, Ling Xu, Fu-Qiang Zhang
ABSTRACT
The TiO2 nanotubes by anodization have been extensively studied for medical implant and orthopedic applications because of enhancing bone development. In the present study, a new nano-foveolae structure verified by SEM and AFM was prepared by simulating the nanotubes exfoliation from anodized Ti. MC3T3-E1 preosteoblasts were used to investigate the effects of new nanoscale surface on cell spreading, proliferation, and osteogenic gene expression. The new nano-foveolae structure supported preosteoblast better spreading, more filopodiaes shown by SEM (4 h), and lower proliferation (72 h) than the smooth Ti. By two weeks, the new nanoscale surface induced higher expression of osteogenic markers alkaline phosphatase (ALP), osteopontin (OPN), osteocalcin (OCN), and collagen I (COL I) with real-time RT-PCR compared to the control group. The results of this study suggest that the new nano-foveolae structure provides a favorable surface for functions of bone cells.
Product Description
Titanium, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, sponge, 1-20 mm, 99.5% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Titanium, rod, 1000mm, diameter 6mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 12.7mm, inside diameter 10.9mm, wall thickness 0.9mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 1000mm, diameter 2mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 2.03mm, inside diameter 1.63mm, wall thickness 0.2mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 9.5mm, inside diameter 8.2mm, wall thickness 0.65mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 0.51mm, inside diameter 0.35mm, wall thickness 0.08mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 3.0mm, inside diameter 1.5mm, wall thickness 0.75mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 6mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 0.81mm, inside diameter 0.61mm, wall thickness 0.1mm, as drawn, 99.6+%
Titanium, tube, 200mm, outside diameter 12.7mm, inside diameter 10.9mm, wall thickness 0.9mm, annealed, 99.6+%
Titanium, rod, 500mm, diameter 16mm, annealed, 99.6+%
Titanium, rod, 1000mm, diameter 2mm, as drawn, 99.99+%
Titanium, wire reel, 500m, diameter 0.25mm, as drawn, 99.6+%
Titanium, wire reel, 500m, diameter 0.25mm, annealed, 99.6+%
Titanium, wire reel, 25m, diameter 0.5mm, as drawn, 99.6+%
Titanium, wire reel, 50m, diameter 0.125mm, annealed, 99.6+%
Titanium, wire reel, 200m, diameter 0.5mm, as drawn, 99.6+%
Titanium, wire reel, 5m, diameter 0.5mm, as drawn, 99.99+%
Titanium, wire reel, 5m, diameter 1.5mm, as drawn, 99.6+%