- Stereospecific Ni-catalyzed cross-coupling of potassium alkenyltrifluoroborates with alkyl halides.
Stereospecific Ni-catalyzed cross-coupling of potassium alkenyltrifluoroborates with alkyl halides.
Organic letters (2014-03-29)
Gary A Molander, O Andreea Argintaru
PMID24666316
ABSTRACT
A general method for the alkenylation of alkyl electrophiles using nearly stoichiometric amounts of the air- and moisture-stable potassium organotrifluoroborates has been developed. Various functional groups were tolerated on both the nucleophilic and electrophilic partner. Reactions of highly substituted E- and Z-alkenyltrifluoroborates, as well as vinyl- and propenyltrifluoroborates, were successful, and no loss of stereochemistry or regiochemistry was observed.
Product Description
Nickel, foil, 2m coil, thickness 0.015mm, 99.9%
Nickel, foil, not light tested, 50x50mm, thickness 0.0025mm, 99.95%
Nickel, rod, 1000mm, diameter 20.0mm, 99%
Nickel, foil, 50x50mm, thickness 0.125mm, as rolled, 99.999%
Nickel, rod, 1000mm, diameter 6.35mm, 99%
Nickel, foil, 25x25mm, thickness 0.3mm, annealed, 99%
Nickel, mesh, 30x30mm, nominal aperture 0.04mm, thickness 0.004mm, wire diameter 0.011mm, 500 wires/inch, open area 60%, electro-formed mesh, 99.9%
Nickel, foil, 50x50mm, thickness 0.3mm, as rolled, 99.999%
Nickel, foil, 25x25mm, thickness 2.0mm, annealed, 99%
Nickel, tube, 500mm, outside diameter 1.8mm, inside diameter 1.6mm, wall thickness 0.1mm, hard, 99.5%
Nickel, pellets, 50g, max. size 20mm, 99.99+%
Nickel, foil, 25x25mm, thickness 0.125mm, as rolled, 99.999%
Nickel, foil, 2m coil, thickness 0.025mm, coil width 600mm, as rolled, 99%
Nickel, foil, 50x50mm, thickness 1.0mm, as rolled, 99.999%
Nickel, mesh, 240x240mm, nominal aperture 0.43mm, thickness 0.004mm, wire diameter 0.067mm, 51 wires/inch, open area 75%, electro-formed mesh, 99.9%
Nickel, foil, 50x50mm, thickness 2.0mm, annealed, 99%
Nickel, foil, 150x300mm, thickness 0.25mm, annealed, 99.98%
Nickel, tube, 500mm, outside diameter 1.5mm, inside diameter 1.4mm, wall thickness 0.05mm, hard, 99.5%
Nickel, pellets, 50g, max. size 5mm, 99.99+%
Nickel, microfoil, disks, 25mm, thinness 1.0μm, specific density 895.8μg/cm2, permanent mylar 3.5μm support, 99.99+%
Nickel, foil, 2m coil, thickness 0.02mm, 99.9%
Nickel, wire reel, 10m, diameter 0.125mm, hard, 99.98%
Nickel, foil, 150x300mm, thickness 0.5mm, annealed, 99.98%
Nickel, mesh, 30x30mm, nominal aperture 0.43mm, thickness 0.004mm, wire diameter 0.067mm, 51 wires/inch, open area 75%, electro-formed mesh, 99.9%