Merck
  • Effects of lead-spiked sediments on freshwater bivalve, Hyridella australis: linking organism metal exposure-dose-response.
Effects of lead-spiked sediments on freshwater bivalve, Hyridella australis: linking organism metal exposure-dose-response.
Aquatic toxicology (Amsterdam, Netherlands) (2014-03-01)
Chamani P M Marasinghe Wadige, Anne M Taylor, William A Maher, Rodney P Ubrihien, Frank Krikowa
ABSTRACT
Lead entering aquatic ecosystems adsorbs to sediments and has the potential to cause adverse effects on the health of benthic organisms. To evaluate the freshwater bivalve Hyridella australis as a bioindicator for sediment toxicity, their exposure-dose and response to lead contaminated sediments (< 0.01, 205 ± 9 and 419 ± 16 μg/g dry mass) was investigated in laboratory microcosms using 28 day exposures. Despite high concentrations of lead in the sediments, organisms accumulated low concentrations of lead in their tissues after 28 days of exposure (low treatment: 2.2 ± 0.2 μg/g dry mass, high treatment: 4.2 ± 0.1 μg/g dry mass), however, accumulated lead concentrations in lead exposed organisms were two fold (low treatment) and four fold (high treatment) higher than that of unexposed organisms (1.2 ± 0.3 μg/g dry mass). Accumulation of lead by H. australis may have occurred as analogues of calcium and magnesium. Labial palps accumulated significantly more lead than other tissues. Of the lead accumulated in the hepatopancreas, 83%-91% was detoxified and stored in metal rich granules. The proportions and concentrations of lead in this fraction increased with lead exposure, which suggests that lead detoxification pathway plays an important role in metal tolerance of H. australis. The biologically active lead was mainly present in the mitochondrial fraction which increased with lead exposure. Total antioxidant capacity of H. australis significantly decreased while lipid peroxidation and lysosomal membrane destabilation increased with lead exposure. This study showed a clear lead exposure-dose-response relationship and indicates that H. australis would be a good biomonitor for lead in freshwater ecosystems.
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Lead, foil, 2m coil, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.12mm, coil width 25.4mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.1mm, as rolled, 99.95%
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Lead, foil, not light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, 10mm disks, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.03mm, 99.99+%
Lead, foil, 1m coil, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.55mm, as rolled, 100%
Lead, foil, 0.5m coil, thickness 0.05mm, as rolled, 97% (sn 1.5%, sb 1.5%)