Merck
  • An integrated evaluation of molecular marker indices and linear alkylbenzenes (LABs) to measure sewage input in a subtropical estuary (Babitonga Bay, Brazil).
An integrated evaluation of molecular marker indices and linear alkylbenzenes (LABs) to measure sewage input in a subtropical estuary (Babitonga Bay, Brazil).
Environmental pollution (Barking, Essex : 1987) (2014-02-22)
César C Martins, Ana Caroline Cabral, Scheyla C T Barbosa-Cintra, Ana Lúcia L Dauner, Fernanda M Souza
ABSTRACT
Babitonga Bay is a South Atlantic estuary with significant ecological function; it is part of the last remaining areas of mangrove communities in the Southern Hemisphere. The aim of this study was to determine the spatial distribution of the faecal sterols and linear alkylbenzenes (LABs) in surface sediments and to perform an integrated evaluation of several molecular marker indices to assess the sewage contamination status in the study area. The highest observed concentrations of faecal sterols (coprostanol + epicoprostanol) and LABs were 6.65 μg g(-1) and 413.3 ng g(-1), respectively. Several faecal sterol indices were calculated and correlated with coprostanol levels; these analyses showed that the index limits presented in the current literature could underestimate the sewage contamination in this study area. For the overall estuarine system, a low sewage impact may be assumed based on the low total mass inventories calculated for coprostanol (between 1.4% and 4.8%).
Sigma-Aldrich
Coprostan-3-ol, ≥98%
Sigma-Aldrich
5α-Cholestan-3β-ol, ≥95%
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Supelco
Benzene, analytical standard
Sigma-Aldrich
5β-Cholestan-3α-ol, ≥95%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Supelco
Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%