- Nanoparticle ejection from Au induced by single Xe ion impacts
Nanoparticle ejection from Au induced by single Xe ion impacts
Physical review letters (2000-12-02)
Birtcher, Donnelly, Schlutig
PMID11102163
ABSTRACT
In situ transmission electron microscopy has been used to observe sputtered Au during Xe ion irradiation in transmission geometry. The sputtered Au was collected on an electron transparent carbon foil. Nanoparticles were observed on the collector foil after they were ejected by single ion impacts. The ejection is from the melt zone formed during the thermal spike phase of a displacement cascade produced near the surface by a single ion impact. Such single ion impacts are also capable of producing craters. Ejected nanoparticles can make a significant contribution to sputtering.
Product Description
Carbon, foil, 150x150mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, foil, 150x150mm, thickness 0.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 150x150mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 100x100mm, thickness 5.0mm, rigid graphite, medium grain size, 99.5%
Carbon, foil, 150x150mm, thickness 5.0mm, rigid graphite, medium grain size, 99.5%
Carbon, foil, 15x15mm, thickness 2.0mm, hOpg
Carbon, foil, 150x150mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 200x200mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 25x25mm, thickness 0.5mm, pyrolytic graphite, 99.99%
Carbon, foil, 500x500mm, thickness 1.0mm, flexible graphite, 99.8%
Carbon, foil, 250x250mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, foil, 500x500mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, foil, 10x10mm, thickness 2.0mm, hOpg
Carbon, foil, 150x150mm, thickness 1.0mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 25x25mm, thickness 0.5mm, rigid graphite, fine grain size, 99.997%
Carbon, foil, 12.5x12.5mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 25x25mm, thickness 0.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 25x25mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 150x150mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, not light tested, 300x300mm, thickness 0.075mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.5mm, rigid graphite, fine grain size, 99.997%
Carbon, foil, 25x25mm, thickness 0.15mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 25x25mm, thickness 2.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 8mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 150x150mm, thickness 0.25mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 250x250mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon, foil, not light tested, 300x300mm, thickness 0.125mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 1.0mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.5mm, rigid graphite, fine grain size, 99.95%