Merck
  • Excess of nonceruloplasmin serum copper in AD correlates with MMSE, CSF [beta]-amyloid, and h-tau.
Excess of nonceruloplasmin serum copper in AD correlates with MMSE, CSF [beta]-amyloid, and h-tau.
Neurology (2006-07-13)
R Squitti, G Barbati, L Rossi, M Ventriglia, G Dal Forno, S Cesaretti, F Moffa, I Caridi, E Cassetta, P Pasqualetti, L Calabrese, D Lupoi, P M Rossini
ABSTRACT
To assess whether serum copper in Alzheimer disease (AD) correlates with cognitive scores, beta-amyloid, and other CSF markers of neurodegeneration. The authors studied copper, ceruloplasmin, total peroxide, and antioxidants levels (TRAP) in serum; beta-amyloid in plasma; and copper, beta-amyloid, h-tau, and P-tau in the CSF of 28 patients with AD and 25 healthy controls, in relation to clinical status. Serum copper (p < 0.0001), peroxides (p = 0.002), a copper fraction unexplained by ceruloplasmin (p < 0.0001), and CSF h-tau (p = 0.001) were increased in AD, whereas serum TRAP (p = 0.03) and CSF beta-amyloid were decreased (p < 0.0001). Plasma beta-amyloid increased with age in healthy controls (r = 0.6; p = 0.05). CSF markers of AD correlated with serum copper variables. CSF copper was partially dependent on the serum copper fraction unexplained by ceruloplasmin (t = 2.2, p = 0.04). CSF beta-amyloid seemed to be related to serum copper (r = -0.46; p = 0.002). Mini-Mental Status Examination scores correlated positively with beta-amyloid (r = 0.46, p = 0.002) and inversely with copper unexplained by ceruloplasmin (r = -0.45, p = 0.003). The authors' results confirm the existence of changes in copper component distribution, particularly the copper fraction unexplained by ceruloplasmin and support the hypothesis of a beta-amyloid and copper connection in Alzheimer disease.
Sigma-Aldrich
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
Copper, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper, shot, −3-+14 mesh, 99%
Copper (S, P), BCR®, certified reference material, chips
Copper, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
Copper, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Copper, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%