Merck
Predicted metabolic drug clearance with increasing adult age.
British journal of clinical pharmacology (2012-08-29)
Thomas M Polasek, Farhaan Patel, Berit P Jensen, Michael J Sorich, Michael D Wiese, Matthew P Doogue
ABSTRACT
To determine the effect of increasing adult age on predicted metabolic drug clearance. Predicted metabolic drug clearances (CLPT ) were determined using in vitro-in vivo extrapolation coupled with physiological-based pharmacokinetic modelling and simulation (IVIVE-PBPK) in Simcyp®. Simulations were conducted using CYP-selective 'probe' drugs with subjects in 5 year age groups (20-25 to 90-95 years). CLPT values were compared with human pharmacokinetic data stratified according to age (young = 20-40 years and elderly = 65-85 years) and gender. Age-related changes in the physiological parameters used for IVIVE of CLPT were described. Predicted metabolic drug clearances decreased with increasing adult age to approximately 65-70 years: caffeine from 1.5 to 1.0 ml min(-1)  kg(-1) (a 33% decrease), S-warfarin from 0.100 to 0.064 ml min(-1)  kg(-1) (36%), S-mephenytoin from 4.1 to 2.5 ml min(-1)  kg(-1) (39%), desipramine from 10.6 to 7.3 ml min(-1)  kg(-1) (31%) and midazolam from 5.4 to 3.9 ml min(-1)  kg(-1) (27%). Except for S-mephenytoin, predictions were within 3.5-fold of clearances from clinical studies when stratified by age and gender. A trend towards higher CLPT was observed in females, but this was only statistically significant in larger virtual trials. Physiological parameters that determine CLPT decreased with increasing adult age: mean microsomal protein g(-1) of liver, liver weight, hepatic blood flow and human serum albumin concentration. Decreased metabolic clearance in the elderly was predicted by Simcyp® and was generally consistent with limited clinical data for four out of five drugs studied and the broader literature for drugs metabolized by CYP enzymes. IVIVE-PBPK may be increasingly useful in predicting metabolic drug clearance in the elderly.
Product Description
Desipramine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Desipramine hydrochloride, ≥98% (TLC), powder
Supelco
Desipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®