Merck
Simple buffers for 3D STORM microscopy.
Biomedical optics express (2013-06-14)
Nicolas Olivier, Debora Keller, Vinoth Sundar Rajan, Pierre Gönczy, Suliana Manley
ABSTRACT
3D STORM is one of the leading methods for super-resolution imaging, with resolution down to 10 nm in the lateral direction, and 30-50 nm in the axial direction. However, there is one important requirement to perform this type of imaging: making dye molecules blink. This usually relies on the utilization of complex buffers, containing different chemicals and sensitive enzymatic systems, limiting the reproducibility of the method. We report here that the commercial mounting medium Vectashield can be used for STORM of Alexa-647, and yields images comparable or superior to those obtained with more complex buffers, especially for 3D imaging. We expect that this advance will promote the versatile utilization of 3D STORM by removing one of its entry barriers, as well as provide a more reproducible way to compare optical setups and data processing algorithms.
Sigma-Aldrich
Phalloidin–Atto 655, suitable for fluorescence, ≥90.0% (sum of isomers, HPLC)
Sigma-Aldrich
Abberior® FLIP 565, maleimide, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Sigma-Aldrich
Atto 655-Streptavidin, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 655, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Atto 655-Biotin, BioReagent, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Atto 655 NHS ester, BioReagent, suitable for fluorescence
Sigma-Aldrich
(±)-α-Lipoic acid, ≥98.0%
Sigma-Aldrich
Atto 655-maleimide, BioReagent, suitable for fluorescence, ≥90% (coupling rate)