Merck
  • Functional and morphological aspects of thallium-induced nephrotoxicity in rats.
Functional and morphological aspects of thallium-induced nephrotoxicity in rats.
Toxicology (1995-02-27)
D Appenroth, S Gambaryan, K Winnefeld, M Leiterer, C Fleck, H Bräunlich
ABSTRACT
Until now the effect of thallium (Tl) on renal function has not been investigated systematically. Therefore, the dose (5, 10, 15, 20 mg Tl2SO4/kg body wt., intraperitoneally) and time-dependence of renal damage was investigated in diuresis experiments on conscious rats. Morphology was evaluated after perfusion fixation in situ. Morphologic changes were localized in the thick ascending limb of the loop of Henle, mostly expressed at the 2nd day after Tl administration, which were completely normalized again at the 10th day. Other parameters such as Tl concentration, changes in water content and the activity of Na+/K(+)-ATPase as well as the diuretic effect of furosemide confirmed the Tl effect to be localized in the renal medulla. One single Tl administration is followed by a decrease in glomerular filtration rate (GFR) and urine volume and an increase of proteinuria. Electrolyte excretion was only slightly changed. All changes were reversible within the 10-day investigation period.
Sigma-Aldrich
Thallium(I) sulfate, ≥99.9% trace metals basis
Sigma-Aldrich
Thallium(I) sulfate, 99.99% trace metals basis