Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Pricing and availability is not currently available.
Titanium, IRMM®, certified reference material, 0.5 mm wire
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Pricing and availability is not currently available.
Titanium, IRMM®, certified reference material, 0.5 mm foil
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Pricing and availability is not currently available.
Titanium, IRMM®, certified reference material, 0.1 mm foil
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Pricing and availability is not currently available.