Merck
  • Perforation of fragment simulating projectiles into goat skin and muscle.
Perforation of fragment simulating projectiles into goat skin and muscle.
Journal of the Royal Army Medical Corps (2013-05-31)
Johno Breeze, G R James, A E Hepper
ABSTRACT
Ballistic gelatin is the most common tissue simulant used to reproduce the penetration of projectiles into muscle but published data to support its use are primarily based on bullets, despite explosive fragments being the most common cause of injury to soldiers on current operational deployments. Published ballistic tests using animal and artificial skin and muscle tissue surrogates also lack standardisation in methodology such that limited comparisons with that of human tissues can currently be made. Three masses of cylindrical NATO standardised fragment simulating projectiles (FSPs) were fired at 20% ballistic gelatin and the hind thighs of a killed goat. Threshold (V(th)) and V(50) velocities required for skin perforation and depth of penetration (DoP) into muscle were compared with gelatin. The intercept and gradient of the linear regression lines for DoP versus velocity were compared between gelatin and goat with significance defined as p<0.05. V(50) goat skin perforation velocities for the 0.16, 0.49 and 1.10 g FSPs were 121.1, 103.7 and 97.8 m/s, respectively. There was a significant difference in the V(50) required to perforate the gelatin surface compared with goat skin for the 0.16 and 0.49 g FSPs but not the 1.10 g. There was no statistical difference in the gradients for DoP versus velocity between animal and gelatin for either the 0.16 or 1.10 g FSPs. This study has produced data for skin perforation velocities and generated algorithms describing velocity versus predicted DoP into muscle for three standardised projectiles, which will be used to improve the fidelity of future injury models. 20% gelatin was demonstrated to accurately reproduce the retardation of the 1.10 g FSPs into goat muscle but the addition of a skin simulant will be required to accurately predict DoP for FSPs less than 1.10 g.
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength
Sigma-Aldrich
Gelatin, tested according to Ph. Eur.
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture