Merck
  • Gender-specific role of HDAC11 in kidney ischemia- and reperfusion-induced PAI-1 expression and injury.
Gender-specific role of HDAC11 in kidney ischemia- and reperfusion-induced PAI-1 expression and injury.
American journal of physiology. Renal physiology (2013-05-10)
Jee In Kim, Kyong-Jin Jung, Hee-Seong Jang, Kwon Moo Park
ABSTRACT
Male gender and the male hormone testosterone increase susceptibility to kidney ischemia and reperfusion (I/R) injury, which is associated with inflammatory responses. Possible involvement of histone deacetylase (HDAC) in inflammatory responses has been suggested. We investigated the gender-specific role of HDACs in plasminogen activator inhibitor type-1 (PAI-1) expression and I/R injury. PAI-1 inhibition protected the kidney from I/R-induced inflammation and functional loss. Among HDACs, only HDAC11 negatively regulated PAI-1 expression in I/R-subjected kidney gender specifically and lipopolysaccharide (LPS)-stimulated mouse monocytes/macrophages. HDAC11 gene silencing increased PAI-1 expression. Chromatin immunoprecipitation assay confirmed binding of HDAC11 to the promoter region of PAI-1 and then release by I/R insult or LPS treatment. I/R-induced HDAC11 release was inhibited by orchiectomy and reversed by dihydrotestosterone treatment. Release of HDAC11 increased acetylation of histone H3. In conclusion, male gender and male hormones accelerate I/R-induced decreases in expression and binding of HDAC11, resulting in an increase in PAI-1 expression. These data provide important insight into gender dimorphism offering HDAC11 as a novel target for I/R injury.
Product Description
Supelco
5α-Dihydrotestosterone (DHT) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard