Merck
Catalytic oxidation of benzene with ozone over Mn/KIT-6.
Journal of nanoscience and nanotechnology (2013-05-08)
Jong Hwa Park, Ji Man Kim, Jongsoo Jurng, Gwi-Nam Bae, Sung Hoon Park, Sang Chai Kim, Jong-Ki Jeon, Young-Kwon Park
ABSTRACT
Benzene is one of the target compounds to be removed from air owing to its carcinogenicity. In this study, benzene oxidation with ozone over a MnOx/KIT-6 catalyst was carried out for the first time. MnOx/KIT-6 was synthesized using two different Mn precursors: Mn acetate and Mn nitrate. The characteristics of the synthesized catalysts were examined by X-ray diffraction, X-ray photoelectron spectroscopy, temperature-programmed reduction, Brunauer-Emmett-Teller (BET) surface area, and N2 adsorption-desorption. The catalytic activity was found to be dependent on the amount of ozone consumed and the dispersion and reducibility of MnOx on the catalyst surface.
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Supelco
Benzene, analytical standard
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Supelco
Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material