Merck
  • Chloride transport-driven alveolar fluid secretion is a major contributor to cardiogenic lung edema.
Chloride transport-driven alveolar fluid secretion is a major contributor to cardiogenic lung edema.
Proceedings of the National Academy of Sciences of the United States of America (2013-05-07)
Esther A Solymosi, Stefanie M Kaestle-Gembardt, István Vadász, Liming Wang, Nils Neye, Cécile Julie Adrienne Chupin, Simon Rozowsky, Ramona Ruehl, Arata Tabuchi, Holger Schulz, Andras Kapus, Rory E Morty, Wolfgang M Kuebler
ABSTRACT
Alveolar fluid clearance driven by active epithelial Na(+) and secondary Cl(-) absorption counteracts edema formation in the intact lung. Recently, we showed that impairment of alveolar fluid clearance because of inhibition of epithelial Na(+) channels (ENaCs) promotes cardiogenic lung edema. Concomitantly, we observed a reversal of alveolar fluid clearance, suggesting that reversed transepithelial ion transport may promote lung edema by driving active alveolar fluid secretion. We, therefore, hypothesized that alveolar ion and fluid secretion may constitute a pathomechanism in lung edema and aimed to identify underlying molecular pathways. In isolated perfused lungs, alveolar fluid clearance and secretion were determined by a double-indicator dilution technique. Transepithelial Cl(-) secretion and alveolar Cl(-) influx were quantified by radionuclide tracing and alveolar Cl(-) imaging, respectively. Elevated hydrostatic pressure induced ouabain-sensitive alveolar fluid secretion that coincided with transepithelial Cl(-) secretion and alveolar Cl(-) influx. Inhibition of either cystic fibrosis transmembrane conductance regulator (CFTR) or Na(+)-K(+)-Cl(-) cotransporters (NKCC) blocked alveolar fluid secretion, and lungs of CFTR(-/-) mice were protected from hydrostatic edema. Inhibition of ENaC by amiloride reproduced alveolar fluid and Cl(-) secretion that were again CFTR-, NKCC-, and Na(+)-K(+)-ATPase-dependent. Our findings show a reversal of transepithelial Cl(-) and fluid flux from absorptive to secretory mode at hydrostatic stress. Alveolar Cl(-) and fluid secretion are triggered by ENaC inhibition and mediated by NKCC and CFTR. Our results characterize an innovative mechanism of cardiogenic edema formation and identify NKCC1 as a unique therapeutic target in cardiogenic lung edema.
Sigma-Aldrich
Amiloride hydrochloride hydrate, ≥98% (HPLC), powder
Supelco
Furosemide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Furosemide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Furosemide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Furosemide