Merck
  • Postextubation pulmonary edema: an unusual cause of transient pulmonary edema.
Postextubation pulmonary edema: an unusual cause of transient pulmonary edema.
JBR-BTR : organe de la Societe royale belge de radiologie (SRBR) = orgaan van de Koninklijke Belgische Vereniging voor Radiologie (KBVR) (2013-04-25)
K Carels, V Herpels, L Cardoen, C Lecluyse, S Traen, J Verschakelen
ABSTRACT
We report a case of sudden onset of respiratory distress caused by pulmonary edema due to laryngospasm. The diagnosis was established by the clinical context and chest X-ray. A CT-scan was performed to narrow down the differential diagnosis and to confirm the diagnosis. Postextubation pulmonary edema due to laryngospasm is a rare entity with a typical clinical and radiographic presentation.
Product Description
Supelco
Furosemide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Furosemide, Pharmaceutical Secondary Standard; Certified Reference Material
Furosemide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Furosemide