Merck
  • On the nature of the light produced within PMMA optical light guides in scintillation fiber-optic dosimetry.
On the nature of the light produced within PMMA optical light guides in scintillation fiber-optic dosimetry.
Physics in medicine and biology (2013-03-09)
François Therriault-Proulx, Luc Beaulieu, Louis Archambault, Sam Beddar
ABSTRACT
The goal of this study was to evaluate the nature of the stem effect light produced within an optical fiber, to quantify its composition, and to evaluate the efficiency of the chromatic technique to remove the stem effect. Spectrometry studies were performed during irradiations of a bare PMMA optical fiber with kilovoltage x-rays from a superficial therapy unit, an Ir-192 high-dose-rate brachytherapy source, a Co-60 external-therapy unit, and megavoltage electrons and x-rays from a linear accelerator. Stem effect spectra can be accurately modeled by a linear combination of the Cerenkov light and fluorescence emitted spectra. Fluorescence light contributes more for lower-energy modalities. Cerenkov light contributes more as the energy increases above the threshold for its production. The chromatic stem effect removal technique is accurate in most of the situations. However, noticeable differences were obtained between very specific high-energy irradiation conditions. It would be advantageous to implement an additional channel in the chromatic stem effect removal chain or implement a spectral approach to independently remove the Cerenkov and the fluorescence components from the signal of interest. This would increase the accuracy and versatility of the actual chromatic stem effect removal technique.
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 20,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 10,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 50,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 100,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 8,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,480,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 4,000