Merck
  • Investigation of isotope dilution mass spectrometric (ID-MS) method to determine niacin in infant formula, breakfast cereals and multivitamins.
Investigation of isotope dilution mass spectrometric (ID-MS) method to determine niacin in infant formula, breakfast cereals and multivitamins.
Food chemistry (2013-02-16)
Hyunju Shin, Byungjoo Kim, Joonhee Lee
ABSTRACT
An isotope dilution LC/mass spectrometric (ID-LC/MS) method was developed as a candidate reference method for the accurate determination of niacin in infant formula, breakfast cereals and multivitamin. After spiking nicotinamide-d(4) as an internal standard, infant formula and breakfast cereal samples were hydrolysed under alkaline condition. Samples were then analysed in SRM mode to detect nicotinic acid and nicotinic acid-d(4) at m/z 124→80 and 127→84, respectively. In the case of multivitamin sample that contains mainly free nicotinamide, LC/MS monitored nicotinamide and nicotinamide-d(4) at their SRM channels of m/z 123→80 and m/z 127→84, respectively, after simple extraction. The repeatability and reproducibility were tested for the validation of the developed ID/LC-MS method. Additionally, the developed analytical method was applied to determine total niacin contents in homogenised infant formula, homogenised multivitamin, and commercially available products including different types of infant formula, breakfast cereals, and multivitamin tablets.
Product Description
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinic acid sodium salt, 98%
Supelco
Niacin (Nicotinic Acid), Pharmaceutical Secondary Standard; Certified Reference Material