Merck
  • Carbohydrate-containing Triton X-100 analogues for membrane protein solubilization and stabilization.
Carbohydrate-containing Triton X-100 analogues for membrane protein solubilization and stabilization.
Molecular bioSystems (2013-02-05)
Pil Seok Chae, Marc J Wander, Kyung Ho Cho, Philip D Laible, Samuel H Gellman
ABSTRACT
Membrane protein manipulation is a challenging task owing to limited tertiary and quaternary structural stability once the protein has been removed from a lipid bilayer. Such instability can be overcome by embedding membrane proteins in detergent micelles formed from amphiphiles with carefully tuned properties. This study introduces a class of easy-to-synthesize amphiphiles, which are designated CGT (Chae's Glyco-Triton) detergents. Some of the agents are well suited for membrane protein solubilization and stabilization.
Sigma-Aldrich
Triton X-305 solution, 70% in H2O
Sigma-Aldrich
Triton X-102
Sigma-Aldrich
Triton X-100 solution, BioUltra, for molecular biology, ~10% in H2O
Octoxinol 10, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Triton X-45
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Triton X-100, peroxide- and carbonyl-free
Sigma-Aldrich
Triton X-100, laboratory grade