Merck
  • One-step dispersion of cellulose nanofibers by mechanochemical esterification in an organic solvent.
One-step dispersion of cellulose nanofibers by mechanochemical esterification in an organic solvent.
ChemSusChem (2012-11-28)
Pei Huang, Min Wu, Shigenori Kuga, Deqian Wang, Dayong Wu, Yong Huang
ABSTRACT
Got a crush: Native cellulose can be dispersed as nanofibers in organic solvents by ball milling with esterification agents. Milling with hexanoyl chloride/DMF gives hexanoyl-coated nanofibers dispersible in several organic solvents. Milling with succinic anhydride/DMSO results water-dispersible nanofibers. The results open the way to new cellulose mechanochemistries.
Product Description
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Hexanoyl chloride, purum, ≥98.0% (GC)
Sigma-Aldrich
Hexanoyl chloride, ≥99%
Sigma-Aldrich
Hexanoyl chloride, 97%