Merck
Manganese: a new contrast agent for lung imaging?
Contrast media & molecular imaging (2012-09-20)
Oliviero L Gobbo, Magdalena Zurek, Frederic Tewes, Carsten Ehrhardt, Yannick Crémillieux
ABSTRACT
Lung parenchyma remains one of the most difficult tissues to be imaged by means of magnetic resonance imaging (MRI). Several MRI techniques are routinely used for lung imaging. However, manganese-enhancement MRI (MEMRI) technique has not been associated with pulmonary MRI. Here, we evaluated T(1) -enhancement in the rat lung after a manganese instillation, using a 4.7 T magnet with a radial ultrashort echo time sequence. Our data showed that the signal intensity was increased in lungs receiving a manganese solution compared with a control solution to the lungs. MR signal enhancements above 30% were measured in lung parenchyma following 200 µl instillation of a 1 mm manganese chloride solution. MEMRI, therefore, may be a useful novel tool for enhancing signal intensity and image contrast in lung tissue.
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Sigma-Aldrich
Manganese(II) chloride, powder and chunks, ≥99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride solution, BioReagent, for molecular biology, storage temp.:room temp
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, meets USP testing specifications
Sigma-Aldrich
Manganese(II) chloride, beads, 98%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride 0.1 M solution