Merck
  • Gallates of isoorientin and (2S)-1,2-propanediol glucoside from the leaves of Schoepfia jasminodora.
Gallates of isoorientin and (2S)-1,2-propanediol glucoside from the leaves of Schoepfia jasminodora.
Journal of natural medicines (2011-03-09)
Yoshiko Tsuboi, Takashi Doi, Katsuyoshi Matsunami, Hideaki Otsuka, Takakazu Shinzato, Yoshio Takeda
ABSTRACT
From the 1-BuOH-soluble fraction of a MeOH extract of the leaves of Schoepfia jasminodora collected from Okinawa Island, two new galloyl esters and 15 known compounds were isolated. The structures of the new compounds were elucidated by spectroscopic analysis to be isoorientin 4″-O-gallate and (2S)-1,2-propanediol 1-O-β-D: -glucopyranoside 6'-O-gallate. The compounds possessing galloyl or cathechol moieties showed equal radical scavenging activity.
Sigma-Aldrich
Isoorientin, ≥98% (HPLC)
Isoorientin, primary reference standard