Merck
  • New insights into the structural characteristics of the arabinogalactan-protein (AGP) fraction of gum arabic.
New insights into the structural characteristics of the arabinogalactan-protein (AGP) fraction of gum arabic.
Journal of agricultural and food chemistry (2008-09-12)
T Mahendran, P A Williams, G O Phillips, S Al-Assaf, T C Baldwin
ABSTRACT
The structural characteristics of the gum exudate of Acacia senegal (gum arabic) have been investigated by monitoring the composition and physicochemical properties before and after treatment with proteolytic enzyme and various alkaline systems. Molecular mass ( M w) and radius of gyration ( R g) measurements were performed using gel permeation chromatography (GPC) coupled to refractive index, UV absorbance, and multiangle light scattering detectors and indicated that the macromolecules present have a compact structure. It was found that treatment with proteolytic enzyme caused the arabinogalactan-protein component (AGP) with average molecular mass approximately 2 x 10 (6) Da to degrade, yielding material of molecular mass approximately 4 x 10 (5) Da, whereas the bulk of the material corresponding to the protein-deficient arabinogalactan component (AG) with molecular mass 4 x 10 (5) remained unaffected. Barium hydroxide was found to hydrolyze the polysaccharide component (AG) itself in addition to the proteinaceous component as demonstrated in control experiments using dextran. However, sodium borohydride/sodium hydroxide treatments were unable to hydrolyze dextran and were assumed to hydrolyze only the proteinaceous component of gum arabic. The AGP component was completely degraded, yielding material of molecular mass approximately 4.5 x 10 (4) Da. It has been concluded, therefore, that the enzyme did not fully hydrolyze all of the protein present and that the AGP component of gum arabic consists of carbohydrate blocks of approximately 4.5 x 10 (4) Da linked to a polypeptide chain consistent with the wattle blossom structure. Because the AGP was degraded to differing extents using a mild and more severe sodium borohydride/sodium hydroxide treatment, it was concluded that the polysaccharide moieties were linked through both O-serine and O-hydroxyproline residues. The gum arabic sample was deglycosylated by treatment with anhydrous hydrogen fluoride and revealed the presence of two putative core proteins of approximately 3 x 10 (4) and approximately 5 x 10 (3) Da, respectively, which correspond to proteins of approximately 250 and 45 amino acids in length. A new model for the structure of the AGP component has been proposed.
Sigma-Aldrich
Barium hydroxide solution, 0.3 N
Sigma-Aldrich
Barium hydroxide octahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Barium hydroxide monohydrate, 98%
Sigma-Aldrich
Barium hydroxide octahydrate, ≥98%
Sigma-Aldrich
Barium hydroxide, technical grade, ~95%