Merck
  • Equilibrium and kinetic studies on the adsorption of aniline compounds from aqueous phase onto bifunctional polymeric adsorbent with sulfonic groups.
Equilibrium and kinetic studies on the adsorption of aniline compounds from aqueous phase onto bifunctional polymeric adsorbent with sulfonic groups.
Chemosphere (2005-10-06)
Cai Jianguo, Li Aimin, Shi Hongyan, Fei Zhenghao, Long Chao, Zhang Quanxing
ABSTRACT
In the present study, a hydrophilic bifunctional polymeric resin (LS-2) with sulfonic groups was synthesized, and the adsorption performance of three aniline compounds, aniline, 4-methylaniline, and 4-nitroaniline onto LS-2 was compared with that on the commercial Amberlite XAD-4. The uptake of the aniline compounds on LS-2 is a procedure of coexistence of physisorption and chemisorption and obeys the pseudo-second order rate equation, while the uptake of the compounds on XAD-4 is merely a physical adsorption and follows the pseudo-first order rate equation. All the isothermal data fit well with the Freundlich model, and the values of K(F) of the compounds adsorbing on LS-2 are much higher than those on XAD-4 suggesting the higher adsorbing capacities on LS-2 than those on XAD-4, which may be attributed to the microporous structure and the polar groups on the network of LS-2 resin. Dynamic adsorption and desorption studies for aniline on LS-2 show that the breakthrough adsorption capacity and the total adsorption capacity are 0.96 and 1.24 mmol per milliliter resin, respectively. Nearly 100% regeneration efficiency for the adsorbent was achieved by 5% hydrochloric acid.
Sigma-Aldrich
N-Methylaniline, purum, ≥98.0% (GC)
Sigma-Aldrich
N-Methylaniline, ≥99%
Sigma-Aldrich
N-Methylaniline, 98%
Supelco
N-Methylaniline, analytical standard