Merck
  • Cadmium-induced apoptosis of primary epithelial lung cells: involvement of Bax and p53, but not of oxidative stress.
Cadmium-induced apoptosis of primary epithelial lung cells: involvement of Bax and p53, but not of oxidative stress.
Cell biology and toxicology (2002-05-07)
M Lag, S Westly, T Lerstad, C Bjørnsrud, M Refsnes, P E Schwarze
ABSTRACT
The lung is a target organ for cadmium (Cd) toxicity. Apoptosis induced by cadmium acetate (CdAc) was studied in alveolar type 2 cells and Clara cells isolated from rat lung. Relatively low concentrations of CdAc (1-10 micromol/L) induced apoptosis after exposure for 20 h. Type 2 cells were more sensitive than Clara cells to Cd-induced apoptosis and loss of cell viability. On exposure to 10 micromol/L CdAc, the levels of the apoptosis-modulating proteins p53 and Bax were increased at 2 h and 5-12 h, respectively. The expression of p53 preceded the expression of Bax and the apoptotic process. The exposure to 10 micromol/L CdAc did not significantly increase the formation of cellular reactive oxygen species (ROS). However, after exposure to a high concentration of CdAc (100 micromol/L), a 30% increase of the ROS level was observed. No significant nitric oxide production was measured following CdAc exposure. Catalase, superoxide dismutase, dimethyl sulfoxide, or tetramethylthiourea did not protect against Cd-induced apoptosis. In conclusion, the results show that Clara cells and type 2 cells are sensitive to Cd-induced apoptosis. Increased levels of p53 and Bax are suggested to be involved in the apoptosis. The apoptosis did not appear to be mediated by oxidative pathways.
Product Description
Cadmium(II) acetate, SAFC Hitech®, anhydrous, 99.995%
Sigma-Aldrich
Cadmium(II) acetate, anhydrous, 99.995%
Sigma-Aldrich
Cadmium acetate dihydrate, reagent grade, 98%
Sigma-Aldrich
Cadmium acetate dihydrate, purum p.a., ≥98.0% (KT)