Merck
  • Quantification of amphetamine plasma concentrations by gas chromatography coupled to mass spectrometry.
Quantification of amphetamine plasma concentrations by gas chromatography coupled to mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2000-03-04)
N Pizarro, J Ortuño, J Segura, M Farré, M Mas, J Camí, R de la Torre
ABSTRACT
We developed a fast and sensitive method for identification and quantification of plasma concentrations of amphetamine using gas chromatography with mass spectrometry detection (GC-MS). Amphetamine-d8 served as internal standard. The method involves a single extraction procedure and an easy treatment of the samples that allowed no losses during the evaporation process. Derivatisation of amphetamine with N-methyl-bis(trifluoroacetamide), a potent acylating agent, provides many advantages to the method compared with common derivatisation reactions usually used for amphetamines. The limits of detection and quantification following this method were 0.43 and 1.42 ng/ml, respectively. The assay has been successfully employed in the quantification of amphetamine in plasma samples from healthy volunteers at four different doses.
Product Description
Supelco
N-Methyl-bis(trifluoroacetamide), LiChropur, ≥99.0% (GC)
Supelco
N-Methyl-bis(trifluoroacetamide), LiChropur, ≥97.0% (GC)