Merck
  • Optimization of a two-step permeabilization fluorescence in situ hybridization (FISH) assay for the detection of Staphylococcus aureus.
Optimization of a two-step permeabilization fluorescence in situ hybridization (FISH) assay for the detection of Staphylococcus aureus.
Journal of clinical laboratory analysis (2011-09-16)
Thomas S Lawson, Russell E Connally, Subramanyam Vemulpad, James A Piper
ABSTRACT
Aspects of the fluorescence in situ hybridization (FISH) method for the detection of clinically important bacteria, such as Staphylococcus aureus, Staphylococcus epidermidis, and Escherichia coli, were investigated for optimization. Various approaches to optimizing the FISH procedure were taken and different methods were compared. To save time, hybridization and washing buffers were prepared beforehand and stored at -20 °C and mixed to their final formamide and NaCl concentrations just before use. The use of 50-ml tubes for hybridization incubation reduced drying out, reagent wastage, and reaction times. A two-step permeabilization FISH assay was developed that used phosphate-buffered saline as a buffer for lysostaphin. It could detect bacteria with DNA probes conjugated to fluorophores with a higher signal intensity and the less expensive biotinylated DNA probes with minimal cell lysis in 1 hr. The two-step assay might be used when the FISH signal is weak, bacterial numbers are low or if there is a need to use other reporter molecules.
Sigma-Aldrich
Lysostaphin from Staphylococcus simulans, recombinant, expressed in E. coli, lyophilized powder
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, BioUltra, ≥97% (SDS-PAGE), Protein 40-60 % by biuret, ≥2,000 units/mg protein
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, aseptically filled
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, lyophilized powder, Protein 50-70 % by biuret, ≥500 units/mg protein