Merck
  • Cooperative sensing of mitochondrial DNA by ZBP1 and cGAS promotes cardiotoxicity.
Cooperative sensing of mitochondrial DNA by ZBP1 and cGAS promotes cardiotoxicity.
Cell (2023-06-24)
Yuanjiu Lei, Jordyn J VanPortfliet, Yi-Fan Chen, Joshua D Bryant, Ying Li, Danielle Fails, Sylvia Torres-Odio, Katherine B Ragan, Jingti Deng, Armaan Mohan, Bing Wang, Olivia N Brahms, Shawn D Yates, Michael Spencer, Carl W Tong, Marcus W Bosenberg, Laura Ciaccia West, Gerald S Shadel, Timothy E Shutt, Jason W Upton, Pingwei Li, A Phillip West
ABSTRACT
Mitochondrial DNA (mtDNA) is a potent agonist of the innate immune system; however, the exact immunostimulatory features of mtDNA and the kinetics of detection by cytosolic nucleic acid sensors remain poorly defined. Here, we show that mitochondrial genome instability promotes Z-form DNA accumulation. Z-DNA binding protein 1 (ZBP1) stabilizes Z-form mtDNA and nucleates a cytosolic complex containing cGAS, RIPK1, and RIPK3 to sustain STAT1 phosphorylation and type I interferon (IFN-I) signaling. Elevated Z-form mtDNA, ZBP1 expression, and IFN-I signaling are observed in cardiomyocytes after exposure to Doxorubicin, a first-line chemotherapeutic agent that induces frequent cardiotoxicity in cancer patients. Strikingly, mice lacking ZBP1 or IFN-I signaling are protected from Doxorubicin-induced cardiotoxicity. Our findings reveal ZBP1 as a cooperative partner for cGAS that sustains IFN-I responses to mitochondrial genome instability and highlight ZBP1 as a potential target in heart failure and other disorders where mtDNA stress contributes to interferon-related pathology.
Sigma-Aldrich
Anti-Tom20/Tomm20 Antibody, clone 2F8.1, clone 2F8.1, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Q-VD-OPh, Non-O-methylated, InSolution, ≥90%, irreversible broad-spectrum inhibitor of caspases
Sigma-Aldrich
2′,3′-Dideoxycytidine, ≥98% (HPLC)
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Anti-DNA Antibody, clone AC-30-10, clone AC-30-10, Chemicon®, from mouse
Sigma-Aldrich
Anti-TFAM Antibody, serum, from rabbit
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Bcl-2 Inhibitor VI, ABT-737, The Bcl-2 Inhibitor VI, ABT-737, also referenced under CAS 852808-04-9, controls the biological activity of Bcl-2. This small molecule/inhibitor is primarily used for Activators/Inducers applications.