Merck
  • Comparison of lifelong and late life exercise on oxidative stress in the cerebellum.
Comparison of lifelong and late life exercise on oxidative stress in the cerebellum.
Neurobiology of aging (2007-11-03)
Li Cui, Tim Hofer, Asha Rani, Christiaan Leeuwenburgh, Thomas C Foster
ABSTRACT
Aging was associated with increased oxidation of DNA, RNA, and lipids in the cerebellum of male rats. DNA and lipid oxidation was reduced by lifelong (94 weeks) voluntary exercise on a running wheel. A reduction in cerebellar lipid oxidation, but not RNA or DNA oxidation, was observed following 3 months of moderate exercise or dietary supplementation of vitamin E, initiated at 18 months of age. The level of lipid oxidation correlated with measures of forelimb grip strength. The results indicate that lifelong exercise attenuates multiple molecular markers of age-related oxidative damage in the cerebellum. In addition, modest exercise initiated late in life can have a beneficial effect on lipid oxidation and motor function.
Sigma-Aldrich
Protein A−Alkaline Phosphatase, protein A from Staphylococcus aureus
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
Avidin–Alkaline Phosphatase, buffered aqueous solution
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material