Merck
  • B cell receptor-induced protein dynamics and the emerging role of SUMOylation revealed by proximity proteomics.
B cell receptor-induced protein dynamics and the emerging role of SUMOylation revealed by proximity proteomics.
Journal of cell science (2023-07-07)
Luqman O Awoniyi, Diogo M Cunha, Alexey V Sarapulov, Sara Hernández-Pérez, Marika Runsala, Blanca Tejeda-González, Vid Šuštar, M Özge Balci, Petar Petrov, Pieta K Mattila
ABSTRACT
Successful B cell activation, which is critical for high-affinity antibody production, is controlled by the B cell antigen receptor (BCR). However, we still lack a comprehensive protein-level view of the very dynamic multi-branched cellular events triggered by antigen binding. Here, we employed APEX2 proximity biotinylation to study antigen-induced changes, 5-15 min after receptor activation, at the vicinity of the plasma membrane lipid rafts, wherein BCR enriches upon activation. The data reveals dynamics of signaling proteins, as well as various players linked to the subsequent processes, such as actin cytoskeleton remodeling and endocytosis. Interestingly, our differential expression analysis identified dynamic responses in various proteins previously not linked to early B cell activation. We demonstrate active SUMOylation at the sites of BCR activation in various conditions and report its functional role in BCR signaling through the AKT and ERK1/2 axes.
Sigma-Aldrich
Anti-α-Tubulin Antibody, clone DM1A, Alexa Fluor 647 conjugate, clone DM1A, from mouse, ALEXA FLUOR 647
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Anti-GOLGA3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Roche
Lysozyme, from hen egg white
Sigma-Aldrich
Anti-α-Tubulin Antibody, clone DM1A, Alexa Fluor 488 conjugate, clone DM1A, Upstate®, from mouse