Merck
  • Epigenetic Control of Translation Checkpoint and Tumor Progression via RUVBL1-EEF1A1 Axis.
Epigenetic Control of Translation Checkpoint and Tumor Progression via RUVBL1-EEF1A1 Axis.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2023-04-20)
Mingli Li, Lu Yang, Anthony K N Chan, Sheela Pangeni Pokharel, Qiao Liu, Nicole Mattson, Xiaobao Xu, Wen-Han Chang, Kazuya Miyashita, Priyanka Singh, Leisi Zhang, Maggie Li, Jun Wu, Jinhui Wang, Bryan Chen, Lai N Chan, Jaewoong Lee, Xu Hannah Zhang, Steven T Rosen, Markus Müschen, Jun Qi, Jianjun Chen, Kevin Hiom, Alexander J R Bishop, Chun-Wei Chen
ABSTRACT
Epigenetic dysregulation is reported in multiple cancers including Ewing sarcoma (EwS). However, the epigenetic networks underlying the maintenance of oncogenic signaling and therapeutic response remain unclear. Using a series of epigenetics- and complex-focused CRISPR screens, RUVBL1, the ATPase component of NuA4 histone acetyltransferase complex, is identified to be essential for EwS tumor progression. Suppression of RUVBL1 leads to attenuated tumor growth, loss of histone H4 acetylation, and ablated MYC signaling. Mechanistically, RUVBL1 controls MYC chromatin binding and modulates the MYC-driven EEF1A1 expression and thus protein synthesis. High-density CRISPR gene body scan pinpoints the critical MYC interacting residue in RUVBL1. Finally, this study reveals the synergism between RUVBL1 suppression and pharmacological inhibition of MYC in EwS xenografts and patient-derived samples. These results indicate that the dynamic interplay between chromatin remodelers, oncogenic transcription factors, and protein translation machinery can provide novel opportunities for combination cancer therapy.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-RUVBL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Anti-acetyl-Histone H4 (Lys8) Antibody, serum, Upstate®