Merck
  • Facilitation of Insulin Effects by Ranolazine in Astrocytes in Primary Culture.
Facilitation of Insulin Effects by Ranolazine in Astrocytes in Primary Culture.
International journal of molecular sciences (2022-10-15)
Adrián Jordá, Martin Aldasoro, Ignacio Campo-Palacio, Jose M Vila, Constanza Aldasoro, Juan Campos-Campos, Carlos Colmena, Sandeep Kumar Singh, Elena Obrador, Soraya L Valles
ABSTRACT
Ranolazine (Rn) is a drug used to treat persistent chronic coronary ischemia. It has also been shown to have therapeutic benefits on the central nervous system and an anti-diabetic effect by lowering blood glucose levels; however, no effects of Rn on cellular sensitivity to insulin (Ins) have been demonstrated yet. The present study aimed to investigate the permissive effects of Rn on the actions of Ins in astrocytes in primary culture. Ins (10-8 M), Rn (10-6 M), and Ins + Rn (10-8 M and 10-6 M, respectively) were added to astrocytes for 24 h. In comparison to control cells, Rn and/or Ins caused modifications in cell viability and proliferation. Rn increased protein expression of Cu/Zn-SOD and the pro-inflammatory protein COX-2 was upregulated by Ins. On the contrary, no significant changes were found in the protein expression of NF-κB and IκB. The presence of Rn produced an increase in p-ERK protein and a significant decrease in COX-2 protein expression. Furthermore, Rn significantly increased the effects of Ins on the expression of p-AKT, p-eNOS, p-ERK, Mn-SOD, and PPAR-γ. In addition, Rn + Ins produced a significant decrease in COX-2 expression. In conclusion, Rn facilitated the effects of insulin on the p-AKT, p-eNOS, p-ERK, Mn-SOD, and PPAR-γ signaling pathways, as well as on the anti-inflammatory and antioxidant effects of the hormone.
Sigma-Aldrich
Anti-PPAR-γ antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-MAP Kinase (ERK-1, ERK-2) antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Anti-phospho-ERK1/2 (pThr202/Tyr204) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (Ser473) Antibody, clone 6F5, clone 6F5, from mouse
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, ~2.0 mg/mL, clone AA2, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-SOD2 antibody produced in mouse, clone GT582, affinity isolated antibody
Sigma-Aldrich
Anti-SOD1 Antibody, clone 6F5, ascites fluid, clone 6F5, from mouse
Sigma-Aldrich
Anti-NFκB Antibody, p65 subunit, active subunit, clone 12H11, clone 12H11, Chemicon®, from mouse
Sigma-Aldrich
Anti-phospho eNOS Antibody (Ser1177), from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-IKB α Antibody, clone 6O8, ZooMAb® Rabbit Monoclonal, recombinant, expressed in HEK 293 cells