Merck
Regulation of ATM and ATR by SMARCAL1 and BRG1.
Biochimica et biophysica acta. Gene regulatory mechanisms (2018-10-15)
Ramesh Sethy, Radhakrishnan Rakesh, Ketki Patne, Vijendra Arya, Tapan Sharma, Dominic Thangminlen Haokip, Reshma Kumari, Rohini Muthuswami
ABSTRACT
The G2/M checkpoint is activated on DNA damage by the ATM and ATR kinases that are regulated by post-translational modifications. In this paper, the transcriptional co-regulation of ATM and ATR by SMARCAL1 and BRG1, both members of the ATP-dependent chromatin remodeling protein family, is described. SMARCAL1 and BRG1 co-localize on the promoters of ATM and ATR; downregulation of SMARCAL1 and BRG1 results in transcriptional repression of ATM/ATR and overriding of the G2/M checkpoint leading to mitotic abnormalities. On doxorubicin-induced DNA damage, SMARCAL1 and BRG1 are upregulated and these two proteins in turn, upregulate the expression of ATM/ATR. The transcriptional response to DNA damage is feedback regulated by phospho-ATM as it binds to the promoters of SMARCAL1, BRG1, ATM and ATR on DNA damage. The regulation of ATM/ATR is rendered non-functional in Schimke Immuno-Osseous Dysplasia where SMARCAL1 is mutated and in Coffin-Siris Syndrome where BRG1 is mutated. Thus, an intricate transcriptional regulation of DNA damage response genes mediated by SMARCAL1 and BRG1 is present in mammalian cells.
Sigma-Aldrich
Anti-phospho-Histone H2AX (pSer139) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Chk2 antibody produced in mouse, clone DCS-270, purified from hybridoma cell culture
Sigma-Aldrich
Anti-phospho-Chk2 (pThr68) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-CDK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-acetyl-Histone H3 (Ac-Lys9) antibody, Mouse monoclonal, clone AH3-120, purified from hybridoma cell culture
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, ~2.0 mg/mL, clone AA2, purified from hybridoma cell culture