Merck
  • Leveraging Compound Promiscuity to Identify Targetable Cysteines within the Kinome.
Leveraging Compound Promiscuity to Identify Targetable Cysteines within the Kinome.
Cell chemical biology (2019-04-16)
Suman Rao, Deepak Gurbani, Guangyan Du, Robert A Everley, Christopher M Browne, Apirat Chaikuad, Li Tan, Martin Schröder, Sudershan Gondi, Scott B Ficarro, Taebo Sim, Nam Doo Kim, Matthew J Berberich, Stefan Knapp, Jarrod A Marto, Kenneth D Westover, Peter K Sorger, Nathanael S Gray
ABSTRACT
Covalent kinase inhibitors, which typically target cysteine residues, represent an important class of clinically relevant compounds. Approximately 215 kinases are known to have potentially targetable cysteines distributed across 18 spatially distinct locations proximal to the ATP-binding pocket. However, only 40 kinases have been covalently targeted, with certain cysteine sites being the primary focus. To address this disparity, we have developed a strategy that combines the use of a multi-targeted acrylamide-modified inhibitor, SM1-71, with a suite of complementary chemoproteomic and cellular approaches to identify additional targetable cysteines. Using this single multi-targeted compound, we successfully identified 23 kinases that are amenable to covalent inhibition including MKNK2, MAP2K1/2/3/4/6/7, GAK, AAK1, BMP2K, MAP3K7, MAPKAPK5, GSK3A/B, MAPK1/3, SRC, YES1, FGFR1, ZAK (MLTK), MAP3K1, LIMK1, and RSK2. The identification of nine of these kinases previously not targeted by a covalent inhibitor increases the number of targetable kinases and highlights opportunities for covalent kinase inhibitor development.
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
5-Iodotubercidin, ≥85%, solid
Sigma-Aldrich
Rosetta(DE3) Competent Cells - Novagen, Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.