Merck
  • The pluripotency-associated gene Dppa4 is dispensable for embryonic stem cell identity and germ cell development but essential for embryogenesis.
The pluripotency-associated gene Dppa4 is dispensable for embryonic stem cell identity and germ cell development but essential for embryogenesis.
Molecular and cellular biology (2009-04-01)
Babita Madan, Vikas Madan, Odile Weber, Philippe Tropel, Carmen Blum, Emmanuelle Kieffer, Stéphane Viville, Hans Jörg Fehling
ABSTRACT
Dppa4 (developmental pluripotency-associated 4) has been identified in several high-profile screens as a gene that is expressed exclusively in pluripotent cells. It encodes a nuclear protein with an SAP-like domain and appears to be associated preferentially with transcriptionally active chromatin. Its exquisite expression pattern and results of RNA interference experiments have led to speculation that Dppa4, as well as its nearby homolog Dppa2, might play essential roles in embryonic stem (ES) cell function and/or germ cell development. To rigorously assess suggested roles, we have generated Dppa4-deficient and Dppa4/Dppa2 doubly deficient ES cells, as well as mice lacking Dppa4. Contrary to predictions, we find that Dppa4 is completely dispensable for ES cell identity and germ cell development. Instead, loss of Dppa4 in mice results in late embryonic/perinatal death and striking skeletal defects with partial penetrance. Thus, surprisingly, Dppa4-deficiency affects tissues that apparently never transcribed the gene, and at least some loss-of-function defects manifest phenotypically at an embryonic stage long after physiologic Dppa4 expression has ceased. Concomitant with targeted gene inactivation, we have introduced into the Dppa4 locus a red fluorescent marker (tandem-dimer red fluorescent protein) that is compatible with green fluorescent proteins and allows noninvasive visualization of pluripotent cells and reprogramming events.
Sigma-Aldrich
B6-White Murine ES Cell Line, The generation of gene-modified mice, created by homologous recombination in embryonic stem (ES) cells, has become a fundamental tool for analyzing gene function.
Sigma-Aldrich
PluriStem 129/S6 Murine ES cells, The PluriStem 129S6 Murine ES cell line is derived from the widely used 129/S6/SvEv strain of mice (at 3.5 days p.c.) & is intended for injection into black or albino host blastocysts.