Merck
  • Biochemical diagnosis of mucopolysaccharidosis in a Mexican reference center.
Biochemical diagnosis of mucopolysaccharidosis in a Mexican reference center.
Genetics and molecular biology (2020-02-28)
Sandra Del Carmen Mendoza-Ruvalcaba, Aniel Jessica Leticia Brambila-Tapia, Jesús Alejandro Juárez-Osuna, Thiago Donizete Da Silva-José, José Elías García-Ortiz
ABSTRACT
Mucopolysaccharidoses (MPS) are a group of genetic disorders, each resulting from the deficiency of one of the lysosomal enzymes that catabolizes mucopolysaccharides. For the accurate diagnosis of the disease, the quantification of a specific enzymatic activity is needed. In the present study, we analyzed seven MPS over several periods of time ranging from 2 to 5 years in a reference center in Mexico. During this time, a total of 761 samples belonging to 505 individuals with suspected MPS were analyzed. A total of 198 (26.01%) positive results were found. Among these, MPS IVA accounted for the highest frequency of positive results (49.10%), followed by MPS III (17.69%, IIIA: 11.80% and IIIB: 5.89%). Adjusting for the number of births per year, the estimated incidence per 100,000 births for MPS analyzed were as follows: MPS I: 0.19, MPS II: 0.15, MPS IIIA: 0.26, MPS IIIB: 0.13, MPS IVA: 1.10, MPS VI: 0.17 and MPS VII: 0.23, and the combined estimated incidence of MPS was 2.23 per 100,000 births; however, this incidence seems to be highly underestimated when compared with the results of newborn screenings.
Sigma-Aldrich
4-Nitrocatechol sulfate dipotassium salt, crystalline