Merck
  • SF3B1 homeostasis is critical for survival and therapeutic response in T cell leukemia.
SF3B1 homeostasis is critical for survival and therapeutic response in T cell leukemia.
Science advances (2022-01-22)
Cuijuan Han, Alireza Khodadadi-Jamayran, Adam H Lorch, Qi Jin, Valentina Serafin, Ping Zhu, Yuliya Politanska, Limin Sun, Blanca T Gutierrez-Diaz, Marina V Pryzhkova, Hiam Abdala-Valencia, Elizabeth Thomas Bartom, Barbara Buldini, Giuseppe Basso, Sadanandan E Velu, Kavitha Sarma, Basil B Mattamana, Byoung-Kyu Cho, Rebecca C Obeng, Young Ah Goo, Philip W Jordan, Aristotelis Tsirigos, Yalu Zhou, Panagiotis Ntziachristos
ABSTRACT
The production of noncanonical mRNA transcripts is associated with cell transformation. Driven by our previous findings on the sensitivity of T cell acute lymphoblastic leukemia (T-ALL) cells to SF3B1 inhibitors, we identified that SF3B1 inhibition blocks T-ALL growth in vivo with no notable associated toxicity. We also revealed protein stabilization of the U2 complex component SF3B1 via deubiquitination. Our studies showed that SF3B1 inhibition perturbs exon skipping, leading to nonsense-mediated decay and diminished levels of DNA damage response-related transcripts, such as the serine/threonine kinase CHEK2, and impaired DNA damage response. We also identified that SF3B1 inhibition leads to a general decrease in R-loop formation. We further demonstrate that clinically used SF3B1 inhibitors synergize with CHEK2 inhibitors and chemotherapeutic drugs to block leukemia growth. Our study provides the proof of principle for posttranslational regulation of splicing components and associated roles and therapeutic implications for the U2 complex in T cell leukemia.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
PR-619, ≥95% (HPLC)
Sigma-Aldrich
ML323, ≥98% (HPLC)
Sigma-Aldrich
Topotecan hydrochloride hydrate, ≥98% (HPLC and enzymatic)
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Chk2 Inhibitor II hydrate, ≥98% (HPLC)