Merck
  • A naturally derived small molecule NDSM253 inhibits IKK1 to suppress inflammation response and promote bone healing after fracture.
A naturally derived small molecule NDSM253 inhibits IKK1 to suppress inflammation response and promote bone healing after fracture.
American journal of translational research (2021-02-03)
Liqi Shen, Yun Xiao, Hui Xie, Hongbin Zhao, Tao Luo, Lin Liu, Xuekun Pan
ABSTRACT
Bone fracture induces an acute inflammatory response in the resident and peripheral monocyte/macrophage cells. Excessive amounts of proinflammatory cytokines can cause severe tissue damage and inhibit bone healing. The proinflammatory cytokine genes are mainly controlled by TLR4/NF-κB (Toll-like receptor 4/Nuclear factor κB). Thus, targeting the molecules in this signaling pathway to decrease the expression of proinflammatory cytokines is an effective strategy to inhibit the inflammatory response. Herein, we identified a naturally derived small molecule NDSM253 that specifically inhibited IKKα (Inhibitor of NF-κB kinase subunit-alpha), a critical component of TLR4/NF-κB signaling. Biochemically, NDMS253 decreased phosphorylation of IκB (Inhibitor of NF-κB), thereby increasing the binding of IκB-NF-κB and suppressing the proinflammatory cytokine gene expression. NDMS253 showed a much stronger inhibitory effect on proinflammatory cytokine gene expression than did the known IKK inhibitors, including ACHP (2-Amino-6-[2-(cyclopropylmethoxy)-6-hydroxyphenyl]-4-(4-piperidinyl)-3-pyridinecarbonitrile), IKK16, and Amlexanox. Administration of these IKK inhibitors in a mouse femoral fracture model showed that NDSM253 suppressed proinflammatory cytokine genes, thereby promoting bone healing, while the other three IKK inhibitors showed a weaker improvement of both bone healing and circulating proinflammatory cytokines. Collectively, our data suggested that NDSM253 might be an effective inhibitor of IKKα that could inhibit inflammatory cytokine action in bone injury.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Xylazine hydrochloride, ≥99.0% (HPLC)
Sigma-Aldrich
Mouse Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-1 β ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-13 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
(±)-Ketamine hydrochloride, solid
Sigma-Aldrich
Mouse Il18 / Interleukin-18 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
RIPA Buffer
Supelco
Buprenorphine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-4 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Mouse IL-15 ELISA Kit, for serum, plasma and cell culture supernatant