Merck
FIP200 restricts RNA virus infection by facilitating RIG-I activation.
Communications biology (2021-07-31)
Lingyan Wang, Kun Song, Wenzhuo Hao, Yakun Wu, Girish Patil, Fang Hua, Yiwen Sun, Chaoqun Huang, Jerry Ritchey, Clinton Jones, Lin Liu, Jun-Lin Guan, Shitao Li
ABSTRACT
Retinoic acid-inducible gene I (RIG-I) senses viral RNA and instigates an innate immune signaling cascade to induce type I interferon expression. Currently, the regulatory mechanisms controlling RIG-I activation remain to be fully elucidated. Here we show that the FAK family kinase-interacting protein of 200 kDa (FIP200) facilitates RIG-I activation. FIP200 deficiency impaired RIG-I signaling and increased host susceptibility to RNA virus infection. In vivo studies further demonstrated FIP200 knockout mice were more susceptible to RNA virus infection due to the reduced innate immune response. Mechanistic studies revealed that FIP200 competed with the helicase domain of RIG-I for interaction with the two tandem caspase activation and recruitment domains (2CARD), thereby facilitating the release of 2CARD from the suppression status. Furthermore, FIP200 formed a dimer and facilitated 2CARD oligomerization, thereby promoting RIG-I activation. Taken together, our study defines FIP200 as an innate immune signaling molecule that positively regulates RIG-I activation.
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from calf thymus, Type I, fibers
Sigma-Aldrich
Anti-FIP200 Antibody, clone 14E11.2, clone 14E11.2, from mouse
Sigma-Aldrich
pT7-FLAG-2 Expression Vector
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green