Merck
  • Compensatory hepatic adaptation accompanies permanent absence of intrahepatic biliary network due to YAP1 loss in liver progenitors.
Compensatory hepatic adaptation accompanies permanent absence of intrahepatic biliary network due to YAP1 loss in liver progenitors.
Cell reports (2021-07-08)
Laura M Molina, Junjie Zhu, Qin Li, Tirthadipa Pradhan-Sundd, Yekaterina Krutsenko, Khaled Sayed, Nathaniel Jenkins, Ravi Vats, Bharat Bhushan, Sungjin Ko, Shikai Hu, Minakshi Poddar, Sucha Singh, Junyan Tao, Prithu Sundd, Aatur Singhi, Simon Watkins, Xiaochao Ma, Panayiotis V Benos, Andrew Feranchak, George Michalopoulos, Kari Nejak-Bowen, Alan Watson, Aaron Bell, Satdarshan P Monga
ABSTRACT
Yes-associated protein 1 (YAP1) regulates cell plasticity during liver injury, regeneration, and cancer, but its role in liver development is unknown. We detect YAP1 activity in biliary cells and in cells at the hepatobiliary bifurcation in single-cell RNA sequencing analysis of developing livers. Deletion of Yap1 in hepatoblasts does not impair Notch-driven SOX9+ ductal plate formation but does prevent the formation of the abutting second layer of SOX9+ ductal cells, blocking the formation of a patent intrahepatic biliary tree. Intriguingly, these mice survive for 8 months with severe cholestatic injury and without hepatocyte-to-biliary transdifferentiation. Ductular reaction in the perihilar region suggests extrahepatic biliary proliferation, likely seeking the missing intrahepatic biliary network. Long-term survival of these mice occurs through hepatocyte adaptation via reduced metabolic and synthetic function, including altered bile acid metabolism and transport. Overall, we show YAP1 as a key regulator of bile duct development while highlighting a profound adaptive capability of hepatocytes.
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Goat Anti-Rat IgG Antibody, biotin-SP conjugate, Species Adsorbed, 1.5 mg/mL, Chemicon®
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Donkey Anti-Rabbit IgG Antibody, biotin-SP conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, biotin-SP conjugate, Species Adsorbed, Chemicon®, from goat