Merck
  • Imine bond transformation of a dynamic Sm(III) macrocycle-based chemosensor: The indirect approach for detecting cyanuric chloride.
Imine bond transformation of a dynamic Sm(III) macrocycle-based chemosensor: The indirect approach for detecting cyanuric chloride.
Analytica chimica acta (2021-01-18)
Kun Zhang, Yan Huang, Yin-Jing Shen, Lin-Feng Zhang, Shuang Ma, Ting-Ting Chen, Ziqiang Zheng, Shishen Zhang, Benxia Li
ABSTRACT
Herein, we report our strategy to develop the efficient chemosensor and real-time monitoring technique for cyanuric chloride (TCT) detection. A luminescent macrocyclic mononuclear Sm(III) complex Sm-2k bearing with two dynamic imine bonds has been constructed via the template synthesis between dialdehyde H2Qk and matched diamine 1,2-bis(2-aminoethoxy)ethane. Sensing experiments reveal that complex Sm-2k exhibits the turn-off fluorescent and colorimetric response for TCT in CH3OH. It is especially encouraging that this optical sensing process is not only rapid within 60 s but also high-efficient in the presence of TCT analogues as well as sensitive with the low limit of detection (LOD, 1.74 μM) and wide linear sensing range. Mechanism studies demonstrate that TCT sensing is mainly based on the imine bond transformation of probe Sm-2k, which is due to the increased acidity induced by TCT. Meanwhile, a smartphone-based analytical method was developed to make complex Sm-2k accessible for the real-time TCT detection by RGB value outputs. It is believed that this work can shed some constructive lights on design of chemosensors and convenient detection technique for highly reactive analytes.
Product Description
Supelco
Atrazine-desethyl-desisopropyl-2-hydroxy, PESTANAL®, analytical standard
Sigma-Aldrich
Cyanuric chloride, 99%
Sigma-Aldrich
Melamine, 99%
Sigma-Aldrich
Tris(2,3-epoxypropyl) isocyanurate
Sigma-Aldrich
3,3-Diphenylpropylamine, 97%
Sigma-Aldrich
6-Methyl-1,3,5-triazine-2,4-diamine, 98%