Merck
  • Substrate promiscuity of the cyclic dipeptide prenyltransferases from Aspergillus fumigatus ( section sign).
Substrate promiscuity of the cyclic dipeptide prenyltransferases from Aspergillus fumigatus ( section sign).
Journal of natural products (2008-12-31)
Huixi Zou, Xiaodong Zheng, Shu-Ming Li
ABSTRACT
This study reports that a series of tryptophan derivatives with modifications on the side chain or at the indole ring were accepted by two cyclic dipeptide prenyltransferases, CdpNPT and FtmPT1, and converted to prenylated derivatives. The structures of the enzymatic products were elucidated by NMR and MS analyses. In comparison to cyclic dipeptides, which were reversely prenylated by CdpNPT at N-1 and in a regular manner by FtmPT1 at C-2, respectively, tryptophan and its simple derivatives were prenylated reversely by both enzymes at N-1. These results demonstrated the substrate promiscuity of both enzymes.
Sigma-Aldrich
5-Fluoro-L-tryptophan, ≥98.0% (HPLC)
Supelco
3-Indoleacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
5-Hydroxy-L-tryptophan, 98% (calc. on dried substance)
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
5-Hydroxy-L-tryptophan, powder
Sigma-Aldrich
3-Indoleacetic acid, 98%
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
α-Methyl-DL-tryptophan, crystalline
Sigma-Aldrich
5-Methyl-DL-tryptophan, tryptophan analog
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Abrine, 99%
Sigma-Aldrich
Tryptamine, ≥97%
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
1-Methyl-DL-tryptophan, 97%
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%