Merck
  • Small molecules as potent biphasic modulators of protein liquid-liquid phase separation.
Small molecules as potent biphasic modulators of protein liquid-liquid phase separation.
Nature communications (2020-11-06)
W Michael Babinchak, Benjamin K Dumm, Sarah Venus, Solomiia Boyko, Andrea A Putnam, Eckhard Jankowsky, Witold K Surewicz
ABSTRACT
Liquid-liquid phase separation (LLPS) of proteins that leads to formation of membrane-less organelles is critical to many biochemical processes in the cell. However, dysregulated LLPS can also facilitate aberrant phase transitions and lead to protein aggregation and disease. Accordingly, there is great interest in identifying small molecules that modulate LLPS. Here, we demonstrate that 4,4'-dianilino-1,1'-binaphthyl-5,5'-disulfonic acid (bis-ANS) and similar compounds are potent biphasic modulators of protein LLPS. Depending on context, bis-ANS can both induce LLPS de novo as well as prevent formation of homotypic liquid droplets. Our study also reveals the mechanisms by which bis-ANS and related compounds modulate LLPS and identify key chemical features of small molecules required for this activity. These findings may provide a foundation for the rational design of small molecule modulators of LLPS with therapeutic value.
Sigma-Aldrich
c-di-AMP sodium salt, ≥98% (HPLC)
Sigma-Aldrich
McCoy′s 5A Medium, Modified, with L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Heparin disaccharide I-H sodium salt
Sigma-Aldrich
poly(A)
Enoxaparin sodium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Maltose solution, BioReagent, for molecular biology, ~20% in H2O
Sigma-Aldrich
Monoclonal Anti-G3BP antibody produced in mouse, clone 2F3, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Sodium benzenesulfonate, 97%
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Anti-Mouse IgG1 (γ1), CF488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Roche
Cell Proliferation Kit I (MTT)