Merck
  • Genome wide gene expression analysis of the posterior capsule in patients with osteoarthritis and knee flexion contracture.
Genome wide gene expression analysis of the posterior capsule in patients with osteoarthritis and knee flexion contracture.
The Journal of rheumatology (2014-10-03)
Thomas Mark Campbell, Guy Trudel, Kayleigh Kristin Wong, Odette Laneuville
ABSTRACT
Knee flexion contractures (KFC) are limitations in the ability to fully extend the knee joint. In people with knee osteoarthritis (OA), KFC are common, impair function, and worsen outcomes after arthroplasty. In KFC, the posterior knee capsule is believed to play a key role, but the pathophysiology remains poorly understood. We sought to identify gene expression differences in the posterior knee capsule of patients with OA with and without KFC. Capsule tissue was obtained from the knees of 12 subjects diagnosed with advanced-stage OA at the time of knee arthroplasty surgery. The presence or absence of KFC allocated patients into 2 groups using a case-control design. Genomewide capsular gene expression was compared between the 2 patient groups. Confirmation of differential expression of the corresponding proteins was performed by immunohistochemistry on tissue sections. There were no significant demographic differences between the patients with OA with KFC and without KFC save for reduced extension in their surgical knee (p<0.01). KFC patients showed a 6.4-fold decrease in CSN1S1 (p=0.017) gene expression and a 3.7-, 2.0-, and 2.6-fold increase in CHAD, Sox9, and Cyr61 gene expression, respectively (p=0.001, 0.004, 0.001, respectively). There were corresponding increases in protein levels for chondroadherin, sex determining region Y-box 9, and casein alphaS1 (all p<0.05). Functional analysis of the differentially expressed genes indicated a strong association with pathways related to the extracellular matrix and to tissue fibrosis. Posterior capsules in endstage OA knees with KFC exhibited differential expression of 4 genes all previously documented to be associated with tissue fibrosis.
Sigma-Aldrich
Anti-CSN1S1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CHAD antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SOX9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution