Merck
  • Glycogen synthase kinase-3β inhibits tubular regeneration in acute kidney injury by a FoxM1-dependent mechanism.
Glycogen synthase kinase-3β inhibits tubular regeneration in acute kidney injury by a FoxM1-dependent mechanism.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-08-20)
Sonali Sinha, Nidhi Dwivedi, James Woodgett, Shixin Tao, Christianna Howard, Timothy A Fields, Abeda Jamadar, Reena Rao, Sonali Sinha, Nidhi Dwivedi, James Woodgett, Shixin Tao, Christianna Howard, Timothy A Fields, Abeda Jamadar, Reena Rao
ABSTRACT
Acute kidney injury (AKI) is characterized by injury to the tubular epithelium that leads to the sudden loss of renal function. Proper tubular regeneration is essential to prevent progression to chronic kidney disease. In this study, we examined the role of FoxM1, a forkhead box family member transcription factor in tubular repair after AKI. Renal FoxM1 expression increased after renal ischemia/reperfusion (I/R)-induced AKI in mouse kidneys. Treatment with thiostrepton, a FoxM1 inhibitor, reduced FoxM1 regulated pro-proliferative factors and cell proliferation in vitro, and tubular regeneration in mouse kidneys after AKI. Glycogen synthase kinase-3 (GSK3) was found to be an upstream regulator of FoxM1 because GSK3 inhibition or renal tubular GSK3β gene deletion significantly increased FoxM1 expression, and improved tubular repair and renal function. GSK3 inactivation increased β-catenin, Cyclin D1, and c-Myc, and reduced cell cycle inhibitors p21 and p27. Importantly, thiostrepton treatment abolished the improved tubular repair in GSK3β knockout mice following AKI. These results demonstrate that FoxM1 is important for renal tubular regeneration following AKI and that GSK3β suppresses tubular repair by inhibiting FoxM1.
Sigma-Aldrich
Thiostrepton, Thiostrepton, CAS 1393-48-2, is an antibiotic that inhibits protein synthesis by preventing binding of GTP to 50S ribosomal subunit.
Sigma-Aldrich
TDZD-8, ≥98% (HPLC), needles
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Roche
5-Bromo-2′-deoxyuridine, >97%, crystalline solid, pkg of 1 g