Merck
  • A Plasma Membrane Nanodomain Ensures Signal Specificity during Osmotic Signaling in Plants.
A Plasma Membrane Nanodomain Ensures Signal Specificity during Osmotic Signaling in Plants.
Current biology : CB (2020-10-10)
Marija Smokvarska, Charbel Francis, Matthieu Pierre Platre, Jean-Bernard Fiche, Carine Alcon, Xavier Dumont, Philippe Nacry, Vincent Bayle, Marcelo Nollmann, Christophe Maurel, Y Jaillais, Alexandre Martiniere
ABSTRACT
In the course of their growth and development, plants have to constantly perceive and react to their environment. This is achieved in cells by the coordination of complex combinatorial signaling networks. However, how signal integration and specificity are achieved in this context is unknown. With a focus on the hyperosmotic stimulus, we use live super-resolution light imaging methods to demonstrate that a Rho GTPase, Rho-of-Plant 6 (ROP6), forms stimuli-dependent nanodomains within the plasma membrane (PM). These nanodomains are necessary and sufficient to transduce production of reactive oxygen species (ROS) that act as secondary messengers and trigger several plant adaptive responses to osmotic constraints. Furthermore, osmotic signal triggers interaction between ROP6 and two NADPH oxidases that subsequently generate ROS. ROP6 nanoclustering is also needed for cell surface auxin signaling, but short-time auxin treatment does not induce ROS accumulation. We show that auxin-induced ROP6 nanodomains, unlike osmotically driven ROP6 clusters, do not recruit the NADPH oxidase, RBOHD. Together, our results suggest that Rho GTPase nano-partitioning at the PM ensures signal specificity downstream of independent stimuli.
Sigma-Aldrich
Bathophenanthrolinedisulfonic acid disodium salt hydrate, 98%
Hamilton® syringe, 7000 series, knurled hub, 7102, volume 2 μL, needle size 23 ga (bevel tip), needle L 70 mm (2.75 in.)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
1-Naphthaleneacetic acid, suitable for plant cell culture, BioReagent, ≥95%, crystalline
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Supelco
Pefabloc® SC, analytical standard
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Murashige and Skoog Basal Medium, powder, suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis