Merck
  • Recent advances of controlled drug delivery using microfluidic platforms.
Recent advances of controlled drug delivery using microfluidic platforms.
Advanced drug delivery reviews (2017-09-19)
Sharma T Sanjay, Wan Zhou, Maowei Dou, Hamed Tavakoli, Lei Ma, Feng Xu, XiuJun Li
ABSTRACT
Conventional systematically-administered drugs distribute evenly throughout the body, get degraded and excreted rapidly while crossing many biological barriers, leaving minimum amounts of the drugs at pathological sites. Controlled drug delivery aims to deliver drugs to the target sites at desired rates and time, thus enhancing the drug efficacy, pharmacokinetics, and bioavailability while maintaining minimal side effects. Due to a number of unique advantages of the recent microfluidic lab-on-a-chip technology, microfluidic lab-on-a-chip has provided unprecedented opportunities for controlled drug delivery. Drugs can be efficiently delivered to the target sites at desired rates in a well-controlled manner by microfluidic platforms via integration, implantation, localization, automation, and precise control of various microdevice parameters. These features accordingly make reproducible, on-demand, and tunable drug delivery become feasible. On-demand self-tuning dynamic drug delivery systems have shown great potential for personalized drug delivery. This review presents an overview of recent advances in controlled drug delivery using microfluidic platforms. The review first briefly introduces microfabrication techniques of microfluidic platforms, followed by detailed descriptions of numerous microfluidic drug delivery systems that have significantly advanced the field of controlled drug delivery. Those microfluidic systems can be separated into four major categories, namely drug carrier-free micro-reservoir-based drug delivery systems, highly integrated carrier-free microfluidic lab-on-a-chip systems, drug carrier-integrated microfluidic systems, and microneedles. Microneedles can be further categorized into five different types, i.e. solid, porous, hollow, coated, and biodegradable microneedles, for controlled transdermal drug delivery. At the end, we discuss current limitations and future prospects of microfluidic platforms for controlled drug delivery.
Sigma-Aldrich
Various Height Reaction Chamber Chip, Fluidic 1099 PS
Sigma-Aldrich
Pearl-chain mixer, Fluidic 658, PC
Sigma-Aldrich
Spiral sorter, Fluidic 382, PMMA
Sigma-Aldrich
Various Height Reaction Chamber Chip, Fluidic 1100 COC
Sigma-Aldrich
Cross-shaped channel chip - 50 μm channel, Fluidic 82, Cyclic Olefin Polymer (COP) with Luer Interface
Sigma-Aldrich
Cross-shaped channel chip - 50 μm channel, Fluidic 82, Cyclic Olefin Copolymer (COC) with Luer Interface
Sigma-Aldrich
Cross-shaped channel chip - 200 μm channel, Fluidic 394, PMMA with Luer Interface
Sigma-Aldrich
Cross-shaped channel chip - 75 μm channel, Fluidic 106, Cyclic Olefin Polymer (COP) with Luer Interface
Sigma-Aldrich
Pearl-chain mixer, Fluidic 658, COC
Sigma-Aldrich
Reaction chamber chip - 20 μl, Fluidic 556, COP
Sigma-Aldrich
Spiral sorter, Fluidic 382, COP
Sigma-Aldrich
Snowman Mixer Chip, Fluidic 1108, COC
Sigma-Aldrich
Male mini luer plug, single, Fluidic 334, PP
Sigma-Aldrich
200 μl tank with Mini Luer interface, Fluidic 926, PP
Sigma-Aldrich
Phase Guide Mixer Chip, Fluidic 533, COC
Sigma-Aldrich
Diffusion mixer chip, Fluidic 186, PC
Sigma-Aldrich
3D-Serpentine Mixer, Fluidic 1079, PC
Sigma-Aldrich
Diffusion mixer chip, Fluidic 186, COP
Sigma-Aldrich
Various Height Reaction Chamber Chip, Fluidic 1100 PS
Sigma-Aldrich
Cross-shaped channel chip - 75 μm channel, Fluidic 106, PMMA with Luer Interface
Sigma-Aldrich
Cross-shaped channel chip - 75 μm channel (double T-junction), Fluidic 202, Cyclic Olefin Copolymer (COC) with Luer Interface
Sigma-Aldrich
Cross-shaped channel chip - 75 μm channel (double T-junction), Fluidic 202, Cyclic Olefin Polymer (COP) with Luer Interface
Sigma-Aldrich
Cross-shaped channel chip - 75 μm channel, Fluidic 106, Cyclic Olefin Copolymer (COP) with Luer Interface
Sigma-Aldrich
Cross-shaped channel chip - 50 μm channel, Fluidic 82, PMMA with Luer Interface
Sigma-Aldrich
Cross-shaped channel chip - 200 μm channel, Fluidic 394, Cyclic Olefin Copolymer (COC) with Luer Interface
Sigma-Aldrich
Cross-shaped channel chip - 200 μm channel, Fluidic 394, Cyclic Olefin Polymer (COP) with Luer Interface
Sigma-Aldrich
Cross-shaped channel chip - 50 μm channel (extended size platform ΙΙ), Fluidic 189, PMMA with Luer Interface
Sigma-Aldrich
Cross-shaped channel chip - 50 μm channel (extended size platform ΙΙ), Fluidic 189, Cyclic Olefin Copolymer (COC) with Luer Interface
Sigma-Aldrich
Male mini luer plug, single, Fluidic 334, TPE
Sigma-Aldrich
Reaction chamber chip - various volume chambers, Fluidic 620, COC