Merck
  • Serotonergic activity-guided phytochemical investigation of the roots of Angelica sinensis.
Serotonergic activity-guided phytochemical investigation of the roots of Angelica sinensis.
Journal of natural products (2006-04-29)
Shixin Deng, Shao-Nong Chen, Ping Yao, Dejan Nikolic, Richard B van Breemen, Judy L Bolton, Harry H S Fong, Norman R Farnsworth, Guido F Pauli
ABSTRACT
Serotonin receptor (5-HT(7)) binding assay-directed fractionation of a methanol extract of the dried roots of Angelica sinensis led to the isolation and identification of 21 compounds including a new phenolic ester, angeliferulate (1), and three new phthalides, 10-angeloylbutylphthalide (2), sinaspirolide (3), and ansaspirolide (4), along with 17 known compounds, p-hydroxyphenethyl trans-ferulate (5), Z-ligustilide (6), Z-butylidenephthalide (7), senkyunolide I (8), Z-6-hydroxy-7-methoxydihydroligustilide (9), N-butylbenzenesulfonamide (10), 11(S),16(R)-dihydroxyoctadeca-9Z,17-diene-12,14-diyn-1-yl acetate (11), (3R,8S)-falcarindiol (12), heptadeca-1-en-9,10-epoxy-4,6-diyne-3,8-diol (13), oplopandiol (14), 8-hydroxy-1-methoxy-, Z-9-heptadecene-4,6-diyn-3-one (15), imperatorin, ferulic acid, vanillin, stigmasterol, sucrose, and 1,3-dilinolenin. This is the first report of a sulfonamide (10) identified from a higher plant source, although its presence needs further investigation. Biosynthetic pathways for dimeric phthalides 3 and 4 are proposed. Compounds 5, 7, 11, 12, 15, and imperatorin exhibited affinity toward 5-HT(7) receptors in a competitive binding assay.
Sigma-Aldrich
Stigmasterol, ~95%
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
trans-Ferulic acid, 99%
Supelco
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)