Merck
  • Flexible linkers in CaMKII control the balance between activating and inhibitory autophosphorylation.
Flexible linkers in CaMKII control the balance between activating and inhibitory autophosphorylation.
eLife (2020-03-10)
Moitrayee Bhattacharyya, Young Kwang Lee, Serena Muratcioglu, Baiyu Qiu, Priya Nyayapati, Howard Schulman, Jay T Groves, John Kuriyan
ABSTRACT
The many variants of human Ca2+/calmodulin-dependent protein kinase II (CaMKII) differ in the lengths and sequences of disordered linkers connecting the kinase domains to the oligomeric hubs of the holoenzyme. CaMKII activity depends on the balance between activating and inhibitory autophosphorylation (on Thr 286 and Thr 305/306, respectively, in the human α isoform). Variation in the linkers could alter transphosphorylation rates within a holoenzyme and the balance of autophosphorylation outcomes. We show, using mammalian cell expression and a single-molecule assay, that the balance of autophosphorylation is flipped between CaMKII variants with longer and shorter linkers. For the principal isoforms in the brain, CaMKII-α, with a ~30 residue linker, readily acquires activating autophosphorylation, while CaMKII-β, with a ~200 residue linker, is biased towards inhibitory autophosphorylation. Our results show how the responsiveness of CaMKII holoenzymes to calcium signals can be tuned by varying the relative levels of isoforms with long and short linkers.
Sigma-Aldrich
Streptavidin from Streptomyces avidinii, recombinant, expressed in E. coli, lyophilized powder
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution